Product Details
- SNP ID
-
rs3691057
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:123398576 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCCTCTTCCACCTTGTTTAACA[G/A]GAACAGTCATGAAGAACTTGCTTCC
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Dpp10
PubMed Links
Gene Details
- Gene
- Dpp10
- Gene Name
- dipeptidylpeptidase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_199021.3 |
1294 |
Missense Mutation |
|
|
NP_950186.3 |
View Full Product Details