Product Details

SNP ID
rs3716722
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:72314811 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTGCTAGAAACCAGGGCTGGGCACA[C/T]AAGAATATTACATTTTGTAATGTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Xrcc5 PubMed Links

Gene Details

Gene
Xrcc5
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009533.2 Intron NP_033559.2

View Full Product Details