Product Details
- SNP ID
-
rs4222516
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:90781641 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAGCATAGAGCACAGACCTGCAA[A/G]GCGTTGATGAGCTGTCGGTCCTCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Col6a3
PubMed Links
Gene Details
- Gene
- Col6a3
- Gene Name
- collagen, type VI, alpha 3
View Full Product Details