Product Details

SNP ID
rs4222579
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:125867976 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCATTGGTGGCGCCTGTAGGCCCAG[C/A]ATTCCATTTTCCTAGATTTCCTAAG
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
Gpr39 PubMed Links

Gene Details

Gene
Gpr39
Gene Name
G protein-coupled receptor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_027677.2 Intron NP_081953.2
Gene
Lypd1
Gene Name
Ly6/Plaur domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145100.3 Intron NP_659568.2

View Full Product Details