Product Details
- SNP ID
-
rs3708411
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:136250579 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAAAAGAAGCTGATGGTGCGGGCC[A/G]TCTTGTTGGAGAGAATCAGACGGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cux1
PubMed Links
Gene Details
- Gene
- Cux1
- Gene Name
- cut-like homeobox 1
There are no transcripts associated with this gene.
- Gene
- Sh2b2
- Gene Name
- SH2B adaptor protein 2
There are no transcripts associated with this gene.
View Full Product Details