Product Details
- SNP ID
-
rs3023064
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:17723652 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGATATTCAGTCTCATTATCATCA[T/G]CAGTTTCCTTCTGGTTCCTATTTAT
- Phenotype
-
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
St7
PubMed Links
Gene Details
- Gene
- St7
- Gene Name
- suppression of tumorigenicity 7
There are no transcripts associated with this gene.
View Full Product Details