Product Details

SNP ID
rs3685213
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:83120793 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAACCAGATGGGATTGCTGTGGATG[A/G]TAAAGGATGGTATCCAGGGCGCGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ino80b PubMed Links

Gene Details

Gene
Ino80b
Gene Name
INO80 complex subunit B
There are no transcripts associated with this gene.

Gene
Mogs
Gene Name
mannosyl-oligosaccharide glucosidase
There are no transcripts associated with this gene.

Gene
Wbp1
Gene Name
WW domain binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083922.1 343 Silent Mutation NP_001077391.1
NM_001083923.1 343 Intron NP_001077392.1
NM_016757.2 343 Silent Mutation NP_058037.2

View Full Product Details