Product Details

SNP ID
rs3688321
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.8:27267288 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTCGCTCAGTGGAGGAACAAGTGTG[T/A]TTGGAACCTGGGTTTAATCCCAAGC
Phenotype
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
Eif4ebp1 PubMed Links

Gene Details

Gene
Eif4ebp1
Gene Name
eukaryotic translation initiation factor 4E binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007918.3 Intron NP_031944.3

View Full Product Details