Product Details

SNP ID
rs3669669
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:13300272 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATCACAGAGCCAGTGGGATCGACTG[A/G]CTTCTTGCTCTGAAATCGCTCGCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Phactr2 PubMed Links

Gene Details

Gene
Phactr2
Gene Name
phosphatase and actin regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033257.4 Intron NP_001028429.1
NM_001195065.1 Intron NP_001181994.1
NM_001195066.1 Intron NP_001181995.1
NM_001195096.1 Intron NP_001182025.1

View Full Product Details