Product Details
- SNP ID
-
rs3668244
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:69747463 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGAAGCCCACATGAAACACAGG[T/C]TTGGCCAATTCGATGTGGCCAAAGT
- Phenotype
-
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Polr2a
PubMed Links
Gene Details
- Gene
- Polr2a
- Gene Name
- polymerase (RNA) II (DNA directed) polypeptide A
There are no transcripts associated with this gene.
View Full Product Details