Product Details

SNP ID
rs4229088
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:99580137 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGTCTGGCCTATTTCTTAGCAGCC[A/G]GAAGTTGGTTAAGGAACTTCCATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Krtap1-4 PubMed Links

Gene Details

Gene
Krtap1-4
Gene Name
keratin associated protein 1-4
There are no transcripts associated with this gene.

Gene
Krtap1-5
Gene Name
keratin associated protein 1-5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_027157.3 880 UTR 3 NP_081433.2

View Full Product Details