Product Details

SNP ID
rs3023766
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:116922731 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCTGTCCCAGCATCTCATCCA[G/T]GAAGGAGGACCTACCGTGTCCTCTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Mgat5b PubMed Links

Gene Details

Gene
Mgat5b
Gene Name
mannoside acetylglucosaminyltransferase 5, isoenzyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172948.3 Intron NP_766536.2

View Full Product Details