Product Details
- SNP ID
-
rs4229720
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:45546279 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCGGGTCAGCTCCTAGAACTCAT[C/T]GCCTCCTGGTGTCAACCAGCCCTTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Atxn1
PubMed Links
Gene Details
- Gene
- Atxn1
- Gene Name
- ataxin 1
There are no transcripts associated with this gene.
- Gene
- Gmpr
- Gene Name
- guanosine monophosphate reductase
There are no transcripts associated with this gene.
View Full Product Details