Product Details
- SNP ID
-
rs3714056
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:68879859 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGGAGGGAGAATTTGGGTGTATC[G/A]CCTGCATATCCACCACACACAGGTT
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Adcy2
PubMed Links
Gene Details
- Gene
- Adcy2
- Gene Name
- adenylate cyclase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153534.2 |
|
Intron |
|
|
NP_705762.2 |
View Full Product Details