Product Details
- SNP ID
-
rs3090591
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:92212253 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATGTACTGCCTAGGAAAATTGAG[A/G]CCTTTACACTCCATTGTCTTTTTCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Msh3
PubMed Links
Gene Details
- Gene
- Msh3
- Gene Name
- mutS homolog 3 (E. coli)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010829.2 |
3573 |
UTR 3 |
|
|
NP_034959.2 |
View Full Product Details