Product Details

SNP ID
rs3088506
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:71521051 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAACTCTCTCATGACCATAAATGTC[A/C]TCAGATGCCATCATCTTTGCAATAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Fam135b PubMed Links

Gene Details

Gene
Fam135b
Gene Name
family with sequence similarity 135, member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177819.3 Intron NP_808487.2

View Full Product Details