Product Details
- SNP ID
-
rs3708161
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:85279253 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTCAGAAGCTGAACACATGAGGC[C/G]TTGCATAAGATCACACAACTGGCCA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Fbln1
PubMed Links
Gene Details
- Gene
- Fbln1
- Gene Name
- fibulin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010180.2 |
|
Intron |
|
|
NP_034310.2 |
View Full Product Details