Product Details
- SNP ID
-
rs13476388
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:25519163 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACAGAGGAGACAGCAGGAGCTTT[C/T]CTATTGGAAAGCTGAGAAGCTCCCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Traf2
PubMed Links
Gene Details
- Gene
- Traf2
- Gene Name
- TNF receptor-associated factor 2
There are no transcripts associated with this gene.
View Full Product Details