Product Details
- SNP ID
-
rs4223251
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:84339280 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCAACCTTTCCTTCAGGCCGCCA[C/T]GGAAAAAGCACAAATTCAATGCCAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Calcrl
PubMed Links
Gene Details
- Gene
- Calcrl
- Gene Name
- calcitonin receptor-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018782.2 |
1573 |
Silent Mutation |
|
|
NP_061252.2 |
View Full Product Details