Product Details
- SNP ID
-
rs3088823
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52167614 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCCATCACTCTGTATCTCTCCT[A/G]GTGGCTGTGTCCTAGAGACTTCAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Nubpl
PubMed Links
Gene Details
- Gene
- Nubpl
- Gene Name
- nucleotide binding protein-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_029760.2 |
|
Intron |
|
|
NP_084036.2 |
View Full Product Details