Product Details
- SNP ID
-
rs3164209
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:96501871 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACATAAAGATAACTAAAGGTTTCT[C/T]GGCATTAGCAATTAACTCAACAACT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pcp4
PubMed Links
Gene Details
- Gene
- Pcp4
- Gene Name
- Purkinje cell protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008791.2 |
|
Intron |
|
|
NP_032817.1 |
View Full Product Details