Product Details
- SNP ID
-
rs4231753
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:21129066 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAAAAACATGAATCTGTGTCCTA[G/T]CCCACCCCTTCCAGCACGCGCTTCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Garem
PubMed Links
Gene Details
- Gene
- Garem
- Gene Name
- GRB2 associated, regulator of MAPK1
View Full Product Details