Product Details

SNP ID
rs3023550
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:136856018 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TATTGCCTTTCTGGGAATTAAATAA[A/T]GTTGTTGTTTTAATTACTGCCAAAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Nr5a2 PubMed Links

Gene Details

Gene
Nr5a2
Gene Name
nuclear receptor subfamily 5, group A, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159769.2 Intron NP_001153241.1
NM_030676.3 Intron NP_109601.1

View Full Product Details