Product Details

SNP ID
rs16324
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:64011689 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTAGTCCATGCTGATCAACCTCCTG[A/G]ATTCCTGACTATAGGTCTGAAAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cox10 PubMed Links

Gene Details

Gene
Cox10
Gene Name
cytochrome c oxidase assembly protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178379.3 Intron NP_848466.1
Gene
Gm12289
Gene Name
predicted gene 12289
There are no transcripts associated with this gene.

View Full Product Details