Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CR1
Entrez Gene ID
102117401
Gene Name
complement C3b/C4b receptor 1 (Knops blood group)
Gene Aliases
-
Chromosome Location
Chr.1: 66077050 - 66309908 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015449302.1 XP_015304788.1 25 - 26 5703 77
XM_015449307.1 XP_015304793.1 25 - 26 5703 77
XM_015449309.1 XP_015304795.1 24 - 25 5561 77
XM_015449313.1 XP_015304799.1 25 - 26 4970 77
XM_015449318.1 XP_015304804.1 18 - 19 4758 77
XM_015449320.1 XP_015304806.1 25 - 26 4971 77

View Full Product Details