Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OXER1
Entrez Gene ID
102147169
Gene Name
oxoeicosanoid receptor 1
Gene Aliases
-
Chromosome Location
Chr.13: 67668603 - 67672521 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005576013.2 XP_005576070.1 1 - 1 2668 64

View Full Product Details