Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM74
Entrez Gene ID
102126466
Gene Name
transmembrane protein 74
Gene Aliases
-
Chromosome Location
Chr.8: 109901685 - 109905814 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563928.2 XP_005563985.1 1 - 2 138 99

View Full Product Details