Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMM2
Entrez Gene ID
102146519
Gene Name
phosphomannomutase 2
Gene Aliases
-
Chromosome Location
Chr.20: 9093323 - 9140092 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285098.1 NP_001272027.1 2 - 3 215 75
XM_015442172.1 XP_015297658.1 2 - 3 261 75
XM_015442173.1 XP_015297659.1 2 - 3 261 75
XM_015442174.1 XP_015297660.1 3 - 4 642 75
XM_015442175.1 XP_015297661.1 3 - 4 340 75
XM_015442177.1 XP_015297663.1 3 - 4 433 75
XM_015442178.1 XP_015297664.1 2 - 3 527 75
XM_015442179.1 XP_015297665.1 2 - 3 306 75
XM_015442180.1 XP_015297666.1 3 - 4 391 75
GenBank mRNA AB125191.1 - 2 - 3 215 75

View Full Product Details