Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THAP7
Entrez Gene ID
102127848
Gene Name
THAP domain containing 7
Gene Aliases
-
Chromosome Location
Chr.10: 30588210 - 30590592 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567910.2 XP_005567967.2 4 - 5 475 72

View Full Product Details