Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASPHD1
Entrez Gene ID
102128287
Gene Name
aspartate beta-hydroxylase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.20: 28389126 - 28394511 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591591.2 XP_005591648.1 1 - 2 1011 64

View Full Product Details