Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRINP3
Entrez Gene ID
102147185
Gene Name
BMP/retinoic acid inducible neural specific 3
Gene Aliases
FAM5C
Chromosome Location
Chr.1: 48114393 - 48514652 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005540259.2 XP_005540316.1 2 - 3 1002 100
XM_005540260.2 XP_005540317.1 2 - 3 1914 100
XM_005540261.2 XP_005540318.1 2 - 3 1641 100

View Full Product Details