Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHST11
Entrez Gene ID
102145281
Gene Name
carbohydrate sulfotransferase 11
Gene Aliases
-
Chromosome Location
Chr.11: 108167819 - 108476047 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015431469.1 XP_015286955.1 1 - 2 1372 90

View Full Product Details