Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS37D
Entrez Gene ID
102144748
Gene Name
VPS37D subunit of ESCRT-I
Gene Aliases
-
Chromosome Location
Chr.3: 48329308 - 48333079 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015446668.1 XP_015302154.1 3 - 4 692 84

View Full Product Details