Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB12
Entrez Gene ID
102136733
Gene Name
zinc finger and BTB domain containing 12
Gene Aliases
-
Chromosome Location
Chr.4: 138953487 - 138956119 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005553450.2 XP_005553507.1 1 - 2 409 121

View Full Product Details