Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB46
Entrez Gene ID
102140622
Gene Name
zinc finger and BTB domain containing 46
Gene Aliases
-
Chromosome Location
Chr.10: 95939390 - 95992797 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015430129.1 XP_015285615.1 2 - 3 1663 76

View Full Product Details