Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GDNF
Entrez Gene ID
102127908
Gene Name
glial cell derived neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.6: 38292589 - 38319853 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015451316.1 XP_015306802.1 1 - 2 1856 142

View Full Product Details