Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALX1
Entrez Gene ID
102139871
Gene Name
ALX homeobox 1
Gene Aliases
-
Chromosome Location
Chr.11: 84572638 - 84594492 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015431293.1 XP_015286779.1 2 - 3 721 87

View Full Product Details