The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005580029.2 | XP_005580086.1 | 1 - 2 | 194 | 55 |
XM_005580030.2 | XP_005580087.1 | 1 - 2 | 192 | 55 |