Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM46
Entrez Gene ID
101864825
Gene Name
RNA binding motif protein 46
Gene Aliases
-
Chromosome Location
Chr.5: 154029376 - 154078996 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005556125.2 XP_005556182.1 4 - 5 1636 92
XM_005556126.2 XP_005556183.1 4 - 5 1524 92
XM_005556127.2 XP_005556184.1 4 - 5 1450 92

View Full Product Details