Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBED8
Entrez Gene ID
102132681
Gene Name
zinc finger BED-type containing 8
Gene Aliases
C6H5orf54
Chromosome Location
Chr.6: 159829422 - 159837325 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558435.2 XP_005558492.1 1 - 2 461 117

View Full Product Details