Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF5
Entrez Gene ID
102142169
Gene Name
Kruppel like factor 5
Gene Aliases
-
Chromosome Location
Chr.17: 55458381 - 55481055 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585997.2 XP_005586054.1 1 - 2 182 145

View Full Product Details