Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK13
Entrez Gene ID
102147127
Gene Name
kallikrein related peptidase 13
Gene Aliases
-
Chromosome Location
Chr.19: 51893336 - 51906581 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590072.2 XP_005590129.1 3 - 4 991 125

View Full Product Details