Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NXPH2
Entrez Gene ID
102129715
Gene Name
neurexophilin 2
Gene Aliases
-
Chromosome Location
Chr.12: 27029937 - 27142375 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005573069.2 XP_005573126.1 2 - 2 743 143

View Full Product Details