Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD18
Entrez Gene ID
102130126
Gene Name
BTB domain containing 18
Gene Aliases
-
Chromosome Location
Chr.14: 15856833 - 15866210 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577836.2 XP_005577893.1 2 - 3 854 71
XM_015434698.1 XP_015290184.1 2 - 3 332 71

View Full Product Details