Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRDM13
Entrez Gene ID
102145038
Gene Name
PR/SET domain 13
Gene Aliases
-
Chromosome Location
Chr.4: 74193663 - 74202537 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552287.1 XP_005552344.1 2 - 3 532 71

View Full Product Details