The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287709.1 | NP_001274638.1 | 4 - 5 | 950 | 93 |
XM_015436106.1 | XP_015291592.1 | 3 - 4 | 958 | 93 | |
GenBank mRNA | AB071099.1 | - | 4 - 5 | 950 | 93 |