The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005594225.2 | XP_005594282.1 | 3 - 3 | 434 | 94 |
XM_005594226.2 | XP_005594283.1 | 2 - 2 | 383 | 94 |