Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPS
Entrez Gene ID
102133112
Gene Name
neuropeptide S
Gene Aliases
-
Chromosome Location
Chr.9: 126942155 - 126945471 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566738.2 XP_005566795.1 2 - 3 102 80

View Full Product Details