Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCL18
Entrez Gene ID
102135120
Gene Name
C-C motif chemokine ligand 18
Gene Aliases
-
Chromosome Location
Chr.16: 32453295 - 32463859 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583498.2 XP_005583555.1 1 - 2 159 89

View Full Product Details