Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102134512
Entrez Gene ID
102134512
Gene Name
C-C motif chemokine 23
Gene Aliases
CCL23
Chromosome Location
Chr.16: 32402809 - 32407756 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583497.2 XP_005583554.1 2 - 3 203 89

View Full Product Details